Learn More
Invitrogen™ ANGPTL3 Polyclonal Antibody

Description
Recommended positive controls: HepG2 conditioned medium. Predicted reactivity: Mouse (80%), Rat (80%), Pig (85%), Bovine (86%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | ANGPTL3 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1.11 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 20% glycerol and 0.025% ProClin 300 |
| Gene | ANGPTL3 |
| Gene Accession No. | Q9Y5C1 |
| Gene Alias | ANG-5; angiopoietin 5; angiopoietin like 3; Angiopoietin-5; angiopoietin-like 3; angiopoietin-like protein 3; Angiopoietin-related protein 3; ANGPT5; ANGPTL3; ANGPTL3(17-221); ANGPTL3(17-224); ANL3; FHBL2; hypl; hypolipidemia; UNQ153/PRO179 |
| Gene Symbols | ANGPTL3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.