Learn More
Invitrogen™ ANGPTL4 Polyclonal Antibody

Description
ANGPTL4 Polyclonal Antibody for Western Blot, IHC (P), Flow
Specifications
Specifications
| Antigen | ANGPTL4 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | ANGPTL4 |
| Gene Accession No. | Q9BY76 |
| Gene Alias | 425O18-1; angiopoietin like 4; angiopoietin-like 4; angiopoietin-like protein 4; Angiopoietin-related protein 4; ANGPTL2; ANGPTL4; ANGPTL4 C-terminal chain; ANGPTL4 N-terminal chain; ARP4; Bk89; Farp; Fasting-i; Fasting-induced adipose factor; FIAF; fibrinogen/angiopoietin-related protein; HARP; hepatic angiopoietin-related protein; hepatic fibrinogen/angiopoietin-related protein; HFARP; major histocompatibility complex region NG27; Ng27; NL2; peroxisome proliferator-activated receptor (PPAR) gamma induced angiopoietin-related protein; PGAR; Pgarg; Pp1158; PPARG angiopoietin related protein; PSEC0166; Secreted protein Bk89; TGQTL; UNQ171; UNQ171/PRO197 |
| Gene Symbols | ANGPTL4 |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.