Learn More
ANGPTL4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP
Rabbit Polyclonal Antibody
Supplier:  Proteintech Group Inc 183741AP150UL
Encompass_Preferred
 View more versions of this product
Catalog No. 50-173-2135
Only null left
Add to Cart
Description
Description
Angiopoietin-like protein 4 (ANGPTL4), is a secreted protein selectively expressed in adipose tissue, liver, and placenta1-3 that plays a variety of roles in vivo, ranging from adipogenesis to angiogenesis to carcinogenesis. The expression of ANGPTL4 is also under nutritional and hormonal control. ANGPTL4 has been observed to induce a strong pro-angiogenic response independent of vascular endothelial growth factor (VEGF), and its expression has been described in hypoxic human tissues as well as a variety of cancers, including liposarcoma, hepatocellular carcinoma, and conventional renal cell carcinoma.
Specifications
Specifications
ANGPTL4
Polyclonal
Unconjugated
ANGPTL4
angiopoietin like 4, Angiopoietin like protein 4, Angiopoietin related protein 4, ANGPTL2, ANGPTL4, ARP4, FIAF, HFARP, NL2, PGAR, pp1158, PSEC0166, UNQ171/PRO197
Rabbit
Antigen Affinity Chromatography
RUO
51129, 57875
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.29 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9BY76, Q9Z1P8
ANGPTL4
ANGPTL4 Fusion Protein Ag13237
150 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.