Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ANGPTL5 Rabbit anti-Human, Polyclonal, FabGennix

Catalog No. 502206060
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-6060 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-6060 Supplier Fabgennix Inc Supplier No. ANGPTL5501AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ANGPTL5 Polyclonal antibody specifically detects ANGPTL5 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot

Specifications

Antigen ANGPTL5
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene ANGPTL5
Gene Accession No. Q86XS5
Gene Alias angiopoietin like 5; angiopoietin-like 5; angiopoietin-like protein 5; Angiopoietin-related protein 5; ANGPTL5; UNQ5795/PRO19600
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 340-388 on human ANGPTL5 protein.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 253935
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.