Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ANGPTL7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA536575
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA536575 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA536575 Supplier Thermo Scientific Supplier No. PA536575
Add to Cart
Add to Cart

ANGPTL7 Polyclonal Antibody

ANGPTL7 Polyclonal Antibody for Western Blot, IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen ANGPTL7
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene ANGPTL7
Gene Accession No. O43827, Q8R1Q3
Gene Alias angiopoietin like 7; angiopoietin-like 7; Angiopoietin-like factor; angiopoietin-like factor (CDT6); angiopoietin-like protein 7; Angiopoietin-related protein 7; ANGPTL7; AngX; CDT6; Cornea-derived transcript 6 protein; dJ647M16.1; UNQ313/PRO356
Gene Symbols ANGPTL7
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 301-346 of Human Angptl7.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10218, 654812
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.