Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Anhydrotetracycline hydrochloride

Catalog No. AAJ66688MB
Click to view available options
Quantity:
10 mg
25 mg

Anhydrotetracycline hydrochloride, CAS # 13803-65-1, is a tetracycline derivative showing antibacterial activity commonly used to control tetracycline-dependent gene expression systems in bacteria. | CAS: 13803-65-1 | C22H23ClN2O6 | 446.88 g/mol

Anhydrotetracycline hydrochloride acts as a useful effector of tetracycline-dependent gene expression in bacterial expression systems.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Anhydrotetracycline hydrochloride is a degradation product derived from tetracycline hydrochloride. It acts as an effector of tetracycline-dependent gene expression in bacterial expression systems. In this system the transcription of genes is reversibly turned on/off in the presence of several compounds including, anhydrotetracycline hydrochloride.
• This compound poorly binds to the 30S ribosomal subunit compared with its precursor, tetracycline.

Applications
• Anhydrotetracycline hydrochloride is commonly used in all fields of biology for control of transcription or translation processes
• This compound has been used in laboratory experiments and shows lower antibacterial activity against Escherichia coli compared with tetracycline.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13803-65-1
Molecular Formula C22H23ClN2O6
Molecular Weight (g/mol) 446.88
MDL Number MFCD00151453
InChI Key FFZXKJVSZDKEMY-XGRJIHFXNA-N
PubChem CID 54710409
IUPAC Name hydrogen (2Z)-2-[amino(hydroxy)methylidene]-4-(dimethylamino)-11,12a-dihydroxy-6-methyl-1,2,3,4,4a,5,12,12a-octahydrotetracene-1,3,12-trione chloride
SMILES [H+].[Cl-].CN(C)C1C2CC3=C(C(O)=C4C=CC=CC4=C3C)C(=O)C2(O)C(=O)\C(=C(\N)O)C1=O

Specifications

Type Anhydrotetracycline Hydrochloride
Physical Form Crystalline Solid
Melting Point 220°C to 223°C
Color Yellow
Quantity 25 mg
Sensitivity Moisture sensitive
Beilstein 4228856
Solubility Information Soluble in ethanol, DMSO, or DMF
Formula Weight 462.88
Chemical Name or Material Anhydrotetracycline hydrochloride
Hazard Category H319-H361d
Precautionary Statement P201-P202-P264b-P280i-P281-P305+P351+P338-P308+P313-P501c
RTECSNumber QI7860000
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C; Store under Argon

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.