Learn More
Aniline, ACS, 99+%
Description
It is used to manufacture dyes, photographic chemicals and agricultural chemicals. Its main use is in the manufacture of precursors to polyurethane and other industrial chemicals. Aniline emulsion template is also essential for the synthesis of hollow microspheres of polyaniline. The poly(N-methyl aniline) doped with sulphonic acids plays an important role for humidity sensors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | °C to 6°C |
| Density | 1.021 |
| Boiling Point | 183°C to 184°C |
| Flash Point | 70°C (158°F) |
| Assay Percent Range | 99+% |
| Refractive Index | 1.586 |
| Quantity | 500 mL |
| Beilstein | 605631 |
| Solubility Information | Slightly soluble in water. |
| Formula Weight | 93.13 |
| Show More |
Safety and Handling
| Hazard Category | H227-H301+H311+H331-H317-H318-H336-H341-H351-H372 |
| Precautionary Statement | P201-P202-P210-P235-P260-P264b-P270-P271-P272-P280g-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P311-P312-P330-P333+P313-P361-P363-P370+P378q-P501c |
| DOTInformation | Hazard Class: 6.1; Packaging Group: II |
| EINECSNumber | 200-539-3 |
| RTECSNumber | BW6650000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.