Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aniline hydrochloride, 99%

Catalog No. AC158410025
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC158410025 2.5 kg Plastic bottle
AC158411000 100 g Plastic bottle
AC158415000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC158410025 Supplier Thermo Scientific Chemicals Supplier No. 158410025
Add to Cart

CAS: 142-04-1 | C6H8ClN | 129.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-04-1
Molecular Formula C6H8ClN
Molecular Weight (g/mol) 129.59
MDL Number MFCD00012958
InChI Key MMCPOSDMTGQNKG-UHFFFAOYSA-N
Synonym aniline hydrochloride, aniline chloride, benzenamine, hydrochloride, anilinium chloride, aniline.hcl, phenylamine hydrochloride, chlorid anilinu, phenylammonium chloride, sul anilinova, chlorhydrate d'aniline
PubChem CID 8870
IUPAC Name aniline;hydrochloride
SMILES [H+].[Cl-].NC1=CC=CC=C1

Specifications

Melting Point 196.0°C to 199.0°C
Color Cream-Yellow to White
Boiling Point 245.0°C
Flash Point 193°C
Loss on Drying 0.5 % max.
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula C6H5NH2·HCl
Quantity 2.5 kg
Merck Index 15,652
Solubility Information Solubility in water: 1070g/L (25°C). Other solubilities: insoluble in ethyl ether and chloroform
Formula Weight 129.59
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Aniline hydrochloride, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Suspected of causing cancer.
Suspected of causing genetic defects.
Toxic if inhaled.
Toxic in contact with skin.
Toxic if swallowed.
Causes serious eye damage.
May cause an allergic skin reaction
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact
EINECSNumber 205-519-8
RTECSNumber CY0875000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.