Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Anillin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584016
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584016 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584016 Supplier Invitrogen™ Supplier No. PA584016
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DLFSDVLEEG ELDMEKSQEE MDQALAESSE EQEDALNISS MSLLAPLAQT VGVVSPESLV STPRLELKDT SRSDESPKPG KFQRTRVPRA ESGDSLGSED RDLLYSIDAY RSQRFKETER PSIKQVIVRK EDV Highest antigen sequence indentity to the following orthologs: Mouse - 86%, Rat - 85%.

This gene encodes an actin-binding protein that plays a role in cell growth and migration, and in cytokinesis. The encoded protein is thought to regulate actin cytoskeletal dynamics in podocytes, components of the glomerulus. Mutations in this gene are associated with focal segmental glomerulosclerosis 8. Alternative splicing results in multiple transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Anillin
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ANLN
Gene Accession No. Q9NQW6
Gene Alias 1110037A17Rik; 2900037I21Rik; actin-binding protein anillin; Anillin; anillin actin binding protein; anillin, actin binding protein; anillin, actin binding protein (scraps homolog, Drosophila); Anln; C78101; DKFZp779A055; FSGS8; RGD1561367; scra; Scraps
Gene Symbols ANLN
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ANLN. Recombinant protein control fragment (Product #RP-103078).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54443
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.