Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Animal Classifying Cards

Fun for the whole family.

Supplier:  Learning Resources, Inc LER2921

 View more versions of this product

Catalog No. S90802B


Includes:

  • Activity card


Description

Description

Kids can learn fun and interesting facts about animals in various habitats.

  • Oversized cards are easy to handle
  • Feature 24 animals in full color on one side
  • Class, habitat, location, size, diet, body covering, and fun facts appear on the other side

Order Info

Grades K-6.

Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.