Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Animal Classifying Cards
Fun for the whole family.
Includes:
- Activity card
| Catalog Number | Mfr. No. | Description | Price | Qty | Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Description | Price | Qty | Availability | ||||
|
S90802A
|
Learning Resources, Inc
LER2920 |
Under the Sea Classifying Cards | Under the Sea Classifying Cards | N/A | N/A | N/A | |||
|
S90802B
|
Learning Resources, Inc
LER2921 |
Animals In the Wild Classifying Cards | Animals In the Wild Classifying Cards | N/A | N/A | N/A | |||
Description
Kids can learn fun and interesting facts about animals in various habitats.
- Oversized cards are easy to handle
- Feature 24 animals in full color on one side
- Class, habitat, location, size, diet, body covering, and fun facts appear on the other side
Order Info
Grades K-6.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title