Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Anisil, 98+%

Catalog No. p-7080864
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 1226-42-2 | C16H14O4 | 270.284 g/mol

Anisil is a biochemical utilized in the preparation of substituted bis(dithiolene)nickel complex. Further, it is used in the preparation of 3-[4,5-bis-(4-methoxy-phenyl)-1H-imidazol-2-yl]-pyridine by reacting with pyridine-3-carbaldehyde.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Anisil is a biochemical utilized in the preparation of substituted bis(dithiolene)nickel complex. Further, it is used in the preparation of 3-[4,5-bis-(4-methoxy-phenyl)-1H-imidazol-2-yl]-pyridine by reacting with pyridine-3-carbaldehyde.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1226-42-2
Molecular Formula C16H14O4
Molecular Weight (g/mol) 270.284
MDL Number MFCD00008405
InChI Key YNANGXWUZWWFKX-UHFFFAOYSA-N
Synonym 4,4'-dimethoxybenzil, anisil, p-anisil, 1,2-bis 4-methoxyphenyl ethane-1,2-dione, bis 4-methoxyphenyl ethanedione, ethanedione, bis 4-methoxyphenyl, di-p-anisoyl, p,p'-dimethoxybenzil, 1,2-ethanedione, 1,2-bis 4-methoxyphenyl, 1,2-bis 4-methoxyphenyl-1,2-ethanedione
PubChem CID 71043
IUPAC Name 1,2-bis(4-methoxyphenyl)ethane-1,2-dione
SMILES COC1=CC=C(C=C1)C(=O)C(=O)C2=CC=C(C=C2)OC

Specifications

Melting Point 132°C to 134°C
Quantity 5 g
Beilstein 531345
Solubility Information Insoluble in water.
Formula Weight 270.29
Percent Purity ≥98%
Chemical Name or Material Anisil
EINECSNumber 214-960-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.