Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Anisoin, 97%

Catalog No. aab2155914
Click to view available options
Quantity:
25 g
500 g

CAS: 119-52-8 | C16H16O4 | 272.30 g/mol

Anisoin is a useful pre-chromatographic derivatization fluorogenic reagent for LC (liquid chromatography) analysis of guanidino compounds. It is widely used as photoinitiators in polymerization reactions. It is widely used in UV curing inks, wood coatings, paper coatings, optical fiber, PCB, screen printing, paper varnish and other surfaces.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Anisoin is a useful pre-chromatographic derivatization fluorogenic reagent for LC (liquid chromatography) analysis of guanidino compounds. It is widely used as photoinitiators in polymerization reactions. It is widely used in UV curing inks, wood coatings, paper coatings, optical fiber, PCB, screen printing, paper varnish and other surfaces.

Solubility
Soluble in water, ethanol and acetone.

Notes
Incompatible with strong oxidizing agents, acid chlorides and acid anhydrides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119-52-8
Molecular Formula C16H16O4
Molecular Weight (g/mol) 272.30
MDL Number MFCD00008411
InChI Key LRRQSCPPOIUNGX-UHFFFAOYNA-N
Synonym anisoin, 2-hydroxy-1,2-bis 4-methoxyphenyl ethanone, 4,4'-dimethoxybenzoin, p-anisoin, ethanone, 2-hydroxy-1,2-bis 4-methoxyphenyl, benzoin, 4,4'-dimethoxy, p,p'-dimethoxybenzoin, benzoin,4'-dimethoxy, 4,4'-anisoin, 1,2-bis 4-methoxyphenyl-2-hydroxyethan-1-one
PubChem CID 95415
IUPAC Name 2-hydroxy-1,2-bis(4-methoxyphenyl)ethanone
SMILES COC1=CC=C(C=C1)C(O)C(=O)C1=CC=C(OC)C=C1

Specifications

Melting Point 110°C to 114°C
Quantity 25 g
Beilstein 3211892
Solubility Information Soluble in water,ethanol and acetone.
Formula Weight 272.3
Percent Purity 97%
Chemical Name or Material Anisoin
EINECSNumber 204-330-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.