Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-Cyclooctene, 95%, stabilized

Catalog No. AC154860025
Click to view available options
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy

Catalog No. AC154860025

Supplier: Thermo Scientific Chemicals 154860025

Only null left
Add to Cart
This item is not returnable. View return policy

CAS: 931-87-3 | C8H14 | 110.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 931-87-3
Molecular Formula C8H14
Molecular Weight (g/mol) 110.20
MDL Number MFCD00001753
InChI Key URYYVOIYTNXXBN-UPHRSURJSA-N
Synonym cis-cyclooctene, z-cyclooctene, cyclooctene, z, cyclooctene, 1z, cyclooct-1-ene, e-cyclooctene, 1-cyclooctene #, 1z-cyclooctene, 7z-cyclooctene, ar,e-cyclooctene
PubChem CID 638079
SMILES C1CCC\C=C/CC1

Specifications

Melting Point -16.0°C
Color Colorless to Brown
Density 0.8400g/mL
Boiling Point 145.0°C to 146.0°C
Flash Point 25°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.4680 to 1.4710
Linear Formula C8H14
Quantity 2.5 L
Specific Gravity 0.84
Solubility Information Solubility in water: <0.1g/L (20°C)
IUPAC Name (Z)-cyclooctene
Stabilizer 100 to 200ppm irganox 1076 FD
Viscosity 1.89 mPa.s (20°C)
Formula Weight 110.2
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material cis-Cyclooctene
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Very toxic to aquatic life with long lasting effects.
Flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Precautionary Statement GHS P Statement
Avoid release to the environment.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Do NOT induce vomiting.
Store in a well
EINECSNumber 213-243-4

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.