Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Anisomycin 97.0+%, TCI America™
SDP

Catalog No. A2613100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A2613100MG 100 mg
A261325MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A2613100MG Supplier TCI America Supplier No. A2613100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 22862-76-6
Molecular Formula C14H19NO4
Molecular Weight (g/mol) 265.31
MDL Number MFCD00077650
InChI Key YKJYKKNCCRKFSL-BFHYXJOUSA-N
Synonym Flagecidin, (2R,3S,4S)-2-(4-Methoxybenzyl)-3,4-pyrrolidinediol 3-Acetate
PubChem CID 253602
ChEBI CHEBI:338412
IUPAC Name (2S,3R,4R)-4-hydroxy-2-[(4-methoxyphenyl)methyl]pyrrolidin-3-yl acetate
SMILES COC1=CC=C(C[C@@H]2NC[C@@H](O)[C@@H]2OC(C)=O)C=C1

Specifications

Type Anisomycin
Physical Form Crystalline Powder
Melting Point 144°C
Color White
Quantity 100 mg
UN Number 3462
Formula Weight 265.31
Percent Purity ≥97.0% (GC)
Chemical Name or Material Anisomycin
RTECSNumber BZ9800000
TSCA No
Recommended Storage Freezer (-20 °C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.