Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ANK2 Mouse anti-Human, FITC, Clone:S105-13, Abnova™

Catalog No. 89333808 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-808 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-808 Supplier Abnova Corporation Supplier No. MAB16998
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptide of human ANK2.

Specifications

Antigen ANK2
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone S105-13
Conjugate FITC
Description ankyrin 2, neuronal
Dilution Immunocytochemistry (1:200)
Immunofluorescence (1:200)
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:200)
Western Blot (1:1000)
The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide)
Gene ANK2
Gene Alias DKFZp686M09125, DKFZp686P0948, FLJ38277, LQT4
Host Species Mouse
Immunogen A synthetic peptide corresponding to amino acids 203-496 of human ANK2.
Quantity 100 μg
Gene ID (Entrez) 287
Target Species Human
Content And Storage Store at -20°C.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.