Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ANKDD1A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546375
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546375 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546375 Supplier Thermo Scientific Supplier No. PA546375
Add to Cart
Add to Cart

ANKDD1A Polyclonal Antibody

ANKDD1A Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen ANKDD1A
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ANKDD1A
Gene Accession No. Q495B1
Gene Alias ANKDD1A; ankyrin repeat and death domain containing 1A; ankyrin repeat and death domain-containing protein 1A
Gene Symbols ANKDD1A
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human ANKDD1A (aa 371-420).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 348094
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.