Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ANKEF1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5121389
This item is not returnable.
View return policy
Description
Antibody tested in Indirect ELISA.

Specifications
ANKEF1 | |
Polyclonal | |
Unconjugated | |
ANKEF1 | |
Ankef1; Ankrd5; ankyrin repeat and EF-hand domain containing 1; ankyrin repeat and EF-hand domain-containing protein 1; ankyrin repeat domain 5; ankyrin repeat domain-containing protein 5; AV276460 | |
Rabbit | |
Protein G | |
RUO | |
63926 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunocytochemistry | |
2.8 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q9NU02 | |
ANKEF1 | |
Recombinant Human Ankyrin repeat and EF-hand domain-containing protein 1 protein (411-698aa). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction