Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ANKFY1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566801
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566801 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566801 Supplier Invitrogen™ Supplier No. PA566801
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ICTRGADMSVP DEKGNPPLWL ALANNLEDIA STLVRHGCDA TCWGPGPGGC LQTLLHRAID ENNEPTACFL IRSGCDVNSP R Highest antigen sequence identity to the following orthologs - mouse 95%, rat 96%.

ANKFY1 encodes a cytoplasmic protein that contains a coiled-coil structure and a BTB/POZ domain at its N-terminus, ankyrin repeats in the middle portion, and a FYVE-finger motif at its C-terminus. This protein belongs to a subgroup of double zinc finger proteins which may be involved in vesicle or protein transport.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANKFY1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ANKFY1
Gene Accession No. Q9P2R3
Gene Alias Ankfy1; ANKHZN; ankyrin repeat and FYVE domain containing 1; ankyrin repeat and FYVE domain-containing protein 1; ankyrin repeat hooked to a zinc finger motif; ankyrin repeat hooked to zinc finger motif; ankyrin repeats hooked to a zinc finger motif; BTBD23; KIAA1255; mKIAA1255; rabankyrin-5; Rank-5; ZFYVE14
Gene Symbols ANKFY1
Host Species Rabbit
Immunogen Recombinant Human ANKFY1. Recombinant protein control fragment (Product #RP-107459).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51479
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.