Learn More
Invitrogen™ ANKH Polyclonal Antibody

Description
This antibody is predicted to react with rat based on sequence homology.
Specifications
Specifications
| Antigen | ANKH |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | ANKH |
| Gene Accession No. | Q9HCJ1, Q9JHZ2 |
| Gene Alias | Ank; ANKH; ANKH inorganic pyrophosphate transport regulator; ankylosis, progressive homolog; CCAL2; CMDJ; CPPDD; D15Ertd221e; FLJ27166; fn54 protein; HANK; KIAA1581; MANK; mKIAA1581; progessive ankylosis-like protein; progressive ankylosis; progressive ankylosis homolog; progressive ankylosis protein; Progressive ankylosis protein homolog; UNQ241/PRO274 |
| Gene Symbols | Ank, ANKH |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.