Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ANKH Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566977
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566977 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566977 Supplier Invitrogen™ Supplier No. PA566977
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ACYVYRKQKKK MENESATEGE DSAMTDMPPT EEVTDIVEMR EENE Highest antigen sequence identity to the following orthologs - mouse 100%, rat 98%.

ANKH is a multipass transmembrane protein that is expressed in joints and other tissues and controls pyrophosphate levels in cultured cells. Progressive ankylosis-mediated control of pyrophosphate levels has been suggested as a possible mechanism regulating tissue calcification and susceptibility to arthritis in higher animals.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANKH
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ANKH
Gene Accession No. Q9HCJ1
Gene Alias Ank; ANKH; ANKH inorganic pyrophosphate transport regulator; ankylosis, progressive homolog; CCAL2; CMDJ; CPPDD; D15Ertd221e; FLJ27166; fn54 protein; HANK; KIAA1581; MANK; mKIAA1581; progessive ankylosis-like protein; progressive ankylosis; progressive ankylosis homolog; progressive ankylosis protein; Progressive ankylosis protein homolog; UNQ241/PRO274
Gene Symbols ANKH
Host Species Rabbit
Immunogen Recombinant Human ANKH. Recombinant protein control fragment (Product #RP-107620).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56172
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.