Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ANKHD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552595
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552595 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552595 Supplier Invitrogen™ Supplier No. PA552595
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QKVERQLQMK TQQQFTKEYL ETKGQKDTVS LHQQCSHRGV FPEGEGDGSL PEDHFSELPQ VDTILFKDND VDDEQQSPPS AEQIDFVPVQ PLSSPQCNFS SDLGSNGTNS LELQKVSGNQ QIVGQPQIAI TGHDQGLLVQ EPD Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 83%.

This gene encodes a protein with multiple ankyrin repeat domains and a single KH-domain. Co-transcription of this gene and the neighboring downstream gene (EIF4EBP3) generates a transcript (MASK-BP3) which encodes a fusion protein comprised of the MASK protein sequence for the majority of the protein and a different C-terminus due to an alternate reading frame for the EIF4EBP3 segments.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANKHD1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ANKHD1
Gene Accession No. Q8IWZ3
Gene Alias 1110004O12Rik; 4933432B13Rik; 9130019P20Rik; A530027J04Rik; A630021B20Rik; AA571404; ANKHD1; ankyrin repeat and KH domain containing 1; ankyrin repeat and KH domain-containing protein 1; HIV-1 Vpr-binding ankyrin repeat protein; hMASK; KIAA1085; MASK; MASK1; mFLJ00246; mKIAA1085; Multiple ankyrin repeats single KH domain; multiple ankyrin repeats, single KH-domain homolog; PP2500; VBARP
Gene Symbols ANKHD1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ANKHD1. Recombinant protein control fragment (Product #RP-102515).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54882
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.