Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ANKIB1 Antibody, Alexa Fluor™ 750, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10061607AF750
Description
ANKIB1 Polyclonal antibody specifically detects ANKIB1 in Human samples. It is validated for Western BlotSpecifications
ANKIB1 | |
Polyclonal | |
Western Blot | |
ankyrin repeat and IBR domain containing 1, ankyrin repeat and IBR domain-containing protein 1, DKFZp434A0225, FLJ33123, KIAA1386DKFZP434A0225 | |
The immunogen recognized by this antibody maps to a region between residue 925 and 975 of human ankyrin repeat and IBR domain-containing protein 1 using the numbering given in entry NP_061877.1 (GeneID 54467). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
54467 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction