Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ANKLE1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567297
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567297 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567297 Supplier Invitrogen™ Supplier No. PA567297
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: STVSDLELLKG LRALGENPHP ITPFTRQLYH QQLEEAQIAP GPEFSGHSLE LAAALRTGCI PDVQADEDAL AQQFEQPDP Highest antigen sequence identity to the following orthologs - mouse 57%, rat 59%.

Endonuclease that probably plays a role in the DNA damage response and DNA repair.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANKLE1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ANKLE1
Gene Accession No. Q8NAG6
Gene Alias 8430438L13Rik; Ankle1; Ankrd41; ankyrin repeat and LEM domain containing 1; ankyrin repeat and LEM domain-containing protein 1; ankyrin repeat domain 41; ankyrin repeat domain-containing protein 41; C430003I23; LEM domain containing 6; Lem3; LEMD6; LEM-domain containing 3; LEM-domain containing protein 3; RGD1308184
Gene Symbols ANKLE1
Host Species Rabbit
Immunogen Recombinant Human ANKLE1. Recombinant protein control fragment (Product #RP-107936).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 126549
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.