Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ANKLE2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551973
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551973 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551973 Supplier Invitrogen™ Supplier No. PA551973
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SETVNKERAN SYKNPRTQDL TAKLRKAVEK GEEDTFSDLI WSNPRYLIGS GDNPTIVQEG CRYNVMHVAA KENQASICQL TLDVLENPDF MRLMYPDDDE AMLQKRIRYV VDLYLNTPDK MGYDTPLHFA CKFGNADVVN VLSSH Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 90%.

This gene encodes a human guanine nucleotide exchange factor. It transduces signals from CRK by binding the SH3 domain of CRK, and activating several members of the Ras family of GTPases. This signaling cascade that may be involved in apoptosis, integrin-mediated signal transduction, and cell transformation. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some variants has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANKLE2
Applications Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ANKLE2
Gene Accession No. Q86XL3
Gene Alias 1110001J12Rik; Ab2-034; AI661024; Ankle2; ankyrin repeat and LEM domain containing 2; ankyrin repeat and LEM domain-containing protein 2; D5Ertd585e; Kiaa0692; LEM domain containing 7; LEM domain-containing protein 4; LEM4; LEMD7; liver regeneration-related protein LRRG057; MCPH16; RGD1310191
Gene Symbols ANKLE2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ANKLE2. Recombinant protein control fragment (Product #RP-102178).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23141
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.