Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ANKLE2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572736
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572736 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572736 Supplier Invitrogen™ Supplier No. PA572736
Add to Cart
Add to Cart

Chicken Polyclonal Antibody

ANKLE2 Polyclonal Antibody for Western Blot, ELISA

This gene encodes a human guanine nucleotide exchange factor. It transduces signals from CRK by binding the SH3 domain of CRK, and activating several members of the Ras family of GTPases. This signaling cascade that may be involved in apoptosis, integrin-mediated signal transduction, and cell transformation. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some variants has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANKLE2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene ANKLE2
Gene Accession No. Q6P1H6, Q7TP65, Q86XL3
Gene Alias 1110001J12Rik; Ab2-034; AI661024; Ankle2; ankyrin repeat and LEM domain containing 2; ankyrin repeat and LEM domain-containing protein 2; D5Ertd585e; Kiaa0692; LEM domain containing 7; LEM domain-containing protein 4; LEM4; LEMD7; liver regeneration-related protein LRRG057; MCPH16; RGD1310191
Gene Symbols ANKLE2
Host Species Chicken
Immunogen ANKLE2 antibody for a 19 amino acid synthetic peptide near the center of human ANKLE2. The immunogen is within amino acids 280 - 330 of ANKLE2.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23141, 360829, 71782
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.