Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ANKMY1 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA550323

Catalog No. PA550323


Only null left
Add to Cart

Description

Description

The antibody detects endogenous levels of total ANKMY1 protein.

ANKMY1 (ankyrin repeat and MYND domain containing 1), also known as ZMYND13 or TSAL1, is a 941 amino acid protein that contains seven ANK repeats, three MORN repeats and one MYND-type zinc finger. MORN repeats were first identified in junctophilins, cytoplasmic proteins involved in junctions between the plasma membrane and the ER/SR membrane. The presence of MORN repeats suggests that ANKMY1 may interact with the plasma membrane. The MYND domain consists of a cluster of cysteine and histidine residues, arranged with an invariant spacing to form a potential zinc-binding motif which may be involved in protein-protein interactions.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ANKMY1
Polyclonal
Unconjugated
ANKMY1
ANKMY1; ankyrin repeat and MYND domain containing 1; ankyrin repeat and MYND domain-containing protein 1; Testis-specific ankyrin-like protein 1; TSAL1; Zinc finger MYND domain-containing protein 13; ZMYND13
Rabbit
Antigen affinity chromatography
RUO
51281
-20°C
Liquid
Immunohistochemistry (Paraffin)
1 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Q9P2S6
ANKMY1
Fusion protein corresponding to residues near the C terminal of human ankyrin repeat and MYND domain containing 1.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.