Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ANKMY1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5140071
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5140071 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5140071 Supplier Invitrogen™ Supplier No. PA5140071
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: PFHTLMPAER ETFLARKRLL EYMGLQLRQA VFAKESQWDP TWLYLCKRAE LIPSHRMKKK GPSLPRGLDV KEQGQIPFFK FCYQCGRSIG VRLLPCPRCY GILTCSKYCK TKAWTEFHKK DCGDLVAIVT QLEQVSRRRE E Highest antigen sequence indentity to the following orthologs: Mouse - 68%, Rat - 65%.

ANKMY1 (ankyrin repeat and MYND domain containing 1), also known as ZMYND13 or TSAL1, is a 941 amino acid protein that contains seven ANK repeats, three MORN repeats and one MYND-type zinc finger. MORN repeats were first identified in junctophilins, cytoplasmic proteins involved in junctions between the plasma membrane and the ER/SR membrane. The presence of MORN repeats suggests that ANKMY1 may interact with the plasma membrane. The MYND domain consists of a cluster of cysteine and histidine residues, arranged with an invariant spacing to form a potential zinc-binding motif which may be involved in protein-protein interactions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANKMY1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ANKMY1
Gene Accession No. Q9P2S6
Gene Alias ANKMY1; ankyrin repeat and MYND domain containing 1; ankyrin repeat and MYND domain-containing protein 1; Testis-specific ankyrin-like protein 1; TSAL1; Zinc finger MYND domain-containing protein 13; ZMYND13
Gene Symbols ANKMY1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ANKMY1. Recombinant protein control fragment (Product #RP-108869).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51281
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.