Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ANKMY2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566894
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566894 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566894 Supplier Invitrogen™ Supplier No. PA566894
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MVHIKKGELTQ EEKELLEVIG KGTVQEAGTL LSSKNVRVNC LDENGMTPLM HAAYKGKLDM CKLLLRHGAD VNCHQHEHGY TALMFAALSG N Highest antigen sequence identity to the following orthologs - mouse 93%, rat 95%.

ANKMY2 (ankyrin repeat and MYND domain containing 2) is a 441 amino acid protein that contains three ANK repeats and one MYND-type zinc finger. Encoded by a gene that maps to human chromosome 7p21.1, ANKMY2 is conserved in chimpanzee, dog, cow, mouse, chicken, zebrafish, fruit fly, mosquito and Caenorhabditis elegans. Downregulation of ANKMY2, associated with frequent deletions of human chromosome 7p22.1, indicate that ANKMY2 may role a role in the pathogenesis of natural killer (NK)-cell malignancies. ANKMY2 is also upregulated by enforced expression of Hox11, which functions broadly to hinder hemopoiesis, diverts differentiation to an alternative fate and promotes pre-leukaemic states.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANKMY2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Ankmy2
Gene Accession No. Q8IV38
Gene Alias AI035571; ANKMY2; ankyrin repeat and MYND domain containing 2; ankyrin repeat and MYND domain-containing protein 2; EGK_13887; Gna14; guanine nucleotide binding protein, alpha 14; LOW QUALITY PROTEIN: ankyrin repeat and MYND domain-containing protein 2; ZMYND20
Gene Symbols Ankmy2
Host Species Rabbit
Immunogen Recombinant Human ANKMY2. Recombinant protein control fragment (Product #RP-105414).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57037
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.