Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ANKMY2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | ANKMY2 |
---|---|
Concentration | 0.27 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-3236
![]() ![]() |
Proteintech Group Inc
210651AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
ANKMY2 (ankyrin repeat and MYND domain containing 2) is a 441 amino acid protein that contains three ANK repeats and one MYND-type zinc finger. Encoded by a gene that maps to human chromosome 7p21.1, ANKMY2 is conserved in chimpanzee, dog, cow, mouse, chicken, zebrafish, fruit fly, mosquito and Caenorhabditis elegans. Downregulation of ANKMY2, associated with frequent deletions of human chromosome 7p22.1, indicate that ANKMY2 may role a role in the pathogenesis of natural killer (NK)-cell maligcies. ANKMY2 is also upregulated by enforced expression of Hox11, which functions broadly to hinder hemopoiesis, diverts differentiation to an alternative fate and promotes pre-leukaemic states.Specifications
ANKMY2 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q3TPE9, Q8IV38 | |
217473, 314046, 57037 | |
ANKMY2 Fusion Protein Ag15185 | |
Primary | |
-20°C | |
ANKMY2 |
0.27 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
ANKMY2, ZMYND20 | |
Ankmy2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title