Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ANKRA2 Antibody (OTI8A9), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272105AF532
Description
ANKRA2 Monoclonal specifically detects ANKRA2 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
ANKRA2 | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
ANKRA2 | |
Full length recombinant protein of human ANKRA2 (NP_075526) produced in E.coli. | |
0.1 mL | |
57763.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI8A9 | |
Western Blot | |
ANKRARFXANK-like protein 2, ankyrin repeat family A protein 2, ankyrin repeat, family A (RFXANK-like), 2 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction