Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ANKRA2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566758
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566758 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566758 Supplier Invitrogen™ Supplier No. PA566758
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MDTSTNLDIGA QLIVEECPST YSLTGMPDIK IEHPLDPNSE EGSAQGVAMG MKFILPNRFD MNVCSRFVKS Highest antigen sequence identity to the following orthologs - mouse 85%, rat 87%.

This protein may facilitate endocytosis by linking megalin to components of the cytoskeleton or endocytic machinery. ANKRA2 is a megalin- and BKCa potassium channel-interacting factor. It is a protein known to interact with HDAC4 and HDAC5.ANKRA2 is important for transcriptional repression by AhRR.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANKRA2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ANKRA2
Gene Accession No. Q9H9E1
Gene Alias 1110004M18Rik; ANKRA; ANKRA2; ankyrin repeat family A member 2; ankyrin repeat family A protein 2; ankyrin repeat family A protein 2-like protein; ankyrin repeat, family A (RFXANK-like), 2; ankyrin-repeat family A protein; RFXANK-like protein 2; similar to SLO-interacting ankyrin-containing protein; SLO-interacting ankyrin-containing protein; wu:fd08c02; zgc:64138
Gene Symbols ANKRA2
Host Species Rabbit
Immunogen Recombinant Human ANKRA2. Recombinant protein control fragment (Product #RP-107415).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57763
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.