Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ ANKRD1/CARP, Mouse, Unlabeled, Clone: 12G8.2,

Catalog No. MABS1228MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABS1228MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS1228MI Supplier MilliporeSigma™ Supplier No. MABS1228
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ankyrin repeat domain-containing protein 1 (UniProt Q15327; also known as Cardiac ankyrin repeat protein, Cytokine-inducible gene C-193 protein, Cytokine-inducible nuclear protein) is encoded by the ANKRD1 (also known as C193, CARP, HA1A2) gene (Gene ID 27063) in human. ANKRD1/CARP belongs to a family of conserved muscle ankyrin repeat proteins (MARPs), members of which also include ANKRD2 and diabetes ankyrin repeat protein (DARP, ANKRD23). ANKRD1 is highly expressed in cardiomyocytes and known to interact with myopalladin and titin, implicating its roles in myopalladin complex-dependent sarcomere integrity maintenance and stress-induced gene regulations medidated by the titin complex. ANKRD1 is up-regulated in patients with dilated cardiomyopathy and in animal models of cardiac hypertrophy induced by pressure overload and adrenergic stimulation. ANKRD1 is shown to regulate the hypertrophic agonist phenylephrine-/PE-induced α-adrenergic signaling pathway in neonatal rat ventricular myocytes, where it recruits and localizes ERK1/2 and GATA4 in a sarcomeric complex, resulting in enhanced GATA4 phosphorylation and subsequent ANKRD1 complex nuclear translocation and gene activation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANKRD1/CARP
Applications Western Blot
Classification Monoclonal
Clone 12G8.2
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene ANKRD1, C193, CARP, HA1A2
Gene Accession No. Q15327
Host Species Mouse
Immunogen GST-tagged recombinant human ANKRD1/CARP fragment corresponding ankyrin repeats 4&5.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_055206
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.