Learn More
ANKRD1 Mouse anti-Human, Mouse, Porcine, Rabbit, Rat, Clone: 1F11F1, ProteinTech

Mouse Monoclonal Antibody
Supplier: Proteintech Group Inc 677751IG150UL

Description
Aliquoting is unnecessary for -20°C storage.
ANKRD1 Monoclonal antibody specifically detects ANKRD1 in Human, Mouse, Porcine, Rabbit, Rat samples. It is validated for Western BlotSpecifications
ANKRD1 | |
Monoclonal | |
1000 μg/mL | |
PBS with 50% glycerol and 0.02% sodium azide | |
Q15327, Q865U8, Q8R560, Q9CR42, Q9TU71 | |
Ankrd1 | |
ANKRD1 Fusion Protein Ag29974 | |
150 μL | |
Primary | |
Human, Mouse, Pig, Rabbit, Rat | |
Antibody | |
IgG2b |
Western Blot | |
1F11F1 | |
Unconjugated | |
Ankrd1 | |
ALRP; ANKRD1; ankyrin repeat domain 1; ankyrin repeat domain 1 (cardiac muscle); ankyrin repeat domain-containing protein 1; ankyrin-like repeat protein; bA320F15.2; C193; C-193; Cardiac adriamycin-responsive protein; cardiac ankyrin repeat protein; cardiac responsive adriamycin protein; Carp; Crap; CVARP; Cytokine-inducible gene C-193 protein; cytokine-inducible nuclear protein; HA1A2; liver ankyrin repeat domain 1; MARP; MARP1; MCARP; muscle ankyrin repeat protein | |
Mouse | |
Protein A | |
RUO | |
100008719, 107765, 27063, 27064, 396959 | |
-20°C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.