Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ANKRD1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988254
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8254 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8254 Supplier Bioss Supplier No. BS8074R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The protein encoded by this gene is localized to the nucleus of endothelial cells and is induced by IL-1 and TNF-alpha stimulation. Studies in rat cardiomyocytes suggest that this gene functions as a transcription factor. Interactions between this protein and the sarcomeric proteins myopalladin and titin suggest that it may also be involved in the myofibrillar stretch-sensor system.

Specifications

Antigen ANKRD1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Ankrd1
Gene Accession No. Q15327
Gene Alias Alrp; Ankrd1; ankyrin repeat domain 1; ankyrin repeat domain 1 (cardiac muscle); Ankyrin repeat domain-containing protein 1; ankyrin-like repeat protein; bA320F15.2; C193; C-193; Cardiac adriamycin-responsive protein; Cardiac ankyrin repeat protein; cardiac responsive adriamycin protein; Carp; Crap; CVARP; Cytokine-inducible gene C-193 protein; Cytokine-inducible nuclear protein; HA1A2; liver ankyrin repeat domain 1; MARP; MARP1; MCARP; muscle ankyrin repeat protein
Gene Symbols Ankrd1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ANKRD1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27063
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.