Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ANKRD10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113415
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113415 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113415 Supplier Invitrogen™ Supplier No. PA5113415
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ANKRD10 Polyclonal Antibody for Western Blot, ELISA

ANKRD10 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANKRD10
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene ANKRD10
Gene Accession No. Q99LW0, Q9NXR5
Gene Alias 4833425P12Rik; ANKRD10; ankyrin repeat domain 10; ankyrin repeat domain-containing protein 10; AW549277
Gene Symbols ANKRD10
Host Species Rabbit
Immunogen Recombinant Human Ankyrin repeat domain-containing protein 10 protein (1-220AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 102334, 55608
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.