Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ANKRD11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565306
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565306 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565306 Supplier Invitrogen™ Supplier No. PA565306
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LSCPSYEEVMH TPRTPSCSAD DYADLVFDCA DSQHSTPVPT APTSACSPSF FDRFSVASSG LSENASQAPA RPLSTNLYRS VSVDIRRTPE EEFSVG Highest antigen sequence identity to the following orthologs - mouse 84%, rat 84%.

ANKRD11 is a member of a novel family of ankyrin repeats containing cofactors (ANCOs) that interact with p160 coactivators to inhibit ligand-dependent transactivation. ANKRD11 encodes a large nuclear protein with five ankyrin repeats, and parts of its sequences have been reported as nasopharyngeal carcinoma susceptibility protein and medulloblastoma antigen. This gene also colocalizes and interacts with histone deacetylases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANKRD11
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ANKRD11
Gene Accession No. Q6UB99
Gene Alias 2410104C19Rik; 3010027A04Rik; 6330578C09Rik; 9530048I21Rik; AA930108; ANCO1; ANCO-1; ANKRD11; ankyrin repeat domain 11; ankyrin repeat domain-containing protein 11; Ankyrin repeat-containing cofactor 1; fc59e05; fi04c06; Gm176; LOW QUALITY PROTEIN: ankyrin repeat domain-containing protein 11; LZ16; nasopharyngeal carcinoma susceptibility protein; T13; truncated ankyrin repeat domain 11 aberrant transcript 1; truncated ankyrin repeat domain 11 aberrant transcript 2; wu:fc59e05; wu:fi04c06; Yod
Gene Symbols ANKRD11
Host Species Rabbit
Immunogen Recombinant Human ANKRD11. Recombinant protein control fragment (Product #RP-106012).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29123
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.