Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ANKRD12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568605
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568605 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568605 Supplier Invitrogen™ Supplier No. PA568605
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 86%; Dog: 86%; Guinea Pig: 86%; Horse: 86%; Human: 86%; Mouse: 93%; Rabbit: 79%; Rat: 93%.

This gene encodes a member of the ankyrin repeats-containing cofactor family. These proteins may inhibit the transcriptional activity of nuclear receptors through the recruitment of histone deacetylases. The encoded protein interacts with p160 coactivators and also represses transcription mediated by the coactivator alteration/deficiency in activation 3 (ADA3). Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANKRD12
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ANKRD12
Gene Accession No. Q6UB98
Gene Alias 2900001A12Rik; AI447928; ANCO1; ANCO2; ANCO-2; Ankrd12; ankyrin repeat domain 12; ankyrin repeat domain-containing 12-like protein; ankyrin repeat domain-containing protein 12; Ankyrin repeat domain-containing protein 12-like protein; ankyrin repeat-containing cofactor 2; AV347965; GAC-1; GAC-1 protein; KIAA0874; mKIAA0874; Nbla00144
Gene Symbols ANKRD12
Host Species Rabbit
Immunogen synthetic peptide directed towards the N-terminal region of human ANKRD12.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23253
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.