Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ANKRD13A Antibody, Alexa Fluor™ 700, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP318496AF700
Description
ANKRD13A Polyclonal antibody specifically detects ANKRD13A in Human, Mouse samples. It is validated for Western BlotSpecifications
ANKRD13A | |
Polyclonal | |
Western Blot | |
ANKRD13, ankyrin repeat domain 13, ankyrin repeat domain 13A, ankyrin repeat domain-containing protein 13A, NY-REN-25, NY-REN-25 antigen, Protein KE03 | |
Recognizes region between residues Between between 500 and 550.. | |
0.1 mL | |
Primary | |
Human, Mouse | |
Purified |
Western Blot | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
88455 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction