Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ANKRD13A Antibody, Novus Biologicals™
SDP

Catalog No. NB396796 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB396796 25 μL
NBP238505 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB396796 Supplier Novus Biologicals Supplier No. NBP23850525UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ANKRD13A Polyclonal specifically detects ANKRD13A in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen ANKRD13A
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.4 μg/mL, Immunohistochemistry, Immunohistochemistry-Paraffin 1:1000 - 1:2500
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Accession No. Q8IZ07
Gene Alias ANKRD13, ankyrin repeat domain 13, ankyrin repeat domain 13A, ankyrin repeat domain-containing protein 13A, NY-REN-25, NY-REN-25 antigen, Protein KE03
Gene Symbols ANKRD13A
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: IQQSLLESSRSQELSGPASNGGISQTNTYDAQYERAIQESLLTSTEGLCPSALSETSRFDNDLQLAMELSAKELEEWELRLQEE
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 88455
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.