Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ANKRD13B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562510
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562510 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562510 Supplier Invitrogen™ Supplier No. PA562510
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: WEALTNSKPG THPMSYEGRR QDRSA Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

ANKRD13B (ankyrin repeat domain 13B) is an ubiquitin-binding protein belongs to the Ankrd 13 family. It specifically recognizes and binds 'Lys-63'-linked ubiquitin, and positively regulates the internalization of ligand-activated EGFR by binding to the Ub moiety of ubiquitinated EGFR at the cell membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANKRD13B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Ankrd13b
Gene Accession No. Q86YJ7
Gene Alias ANKRD13B; ankyrin repeat domain 13B; ankyrin repeat domain-containing protein 13B; AW124583; B930093C12Rik; RGD1564005
Gene Symbols Ankrd13b
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ANKRD13B. Recombinant protein control fragment (Product #RP-101599).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 124930
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.