Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ANKRD13C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572668
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572668 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572668 Supplier Invitrogen™ Supplier No. PA572668
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ANKRD13C Polyclonal Antibody for Western Blot

Acts as a molecular chaperone for G protein-coupled receptors, regulating their biogenesis and exit from the ER.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANKRD13C
Applications Western Blot
Classification Polyclonal
Concentration 1.22 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene ANKRD13C
Gene Accession No. Q7ZUV0
Gene Alias AI505652; Ankrd13c; ankyrin repeat domain 13c; ankyrin repeat domain-containing protein 13C; AU022220; dJ677H15.3; fc57c01; RP4-677H15.5; similar to hypothetical protein DKFZp566D1346; wu:fc57c01; wu:fq56c05; zgc:56077
Gene Symbols ANKRD13C
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 475-508 amino acids of DANRE ankrd13c.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 325167
Target Species Zebrafish
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.