Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ANKRD16 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558167
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558167 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558167 Supplier Invitrogen™ Supplier No. PA558167
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KRCQYEPDYR DNCGVTALMD AIQCGHIDVA RLLLDEHGAC LSAEDSLGAQ ALHRAAVTGQ DEAIRFLVSE LGVDVDVRAT STHLTA Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 77%.

Required to prevent the misactivation of serine (Ser) with tRNA(Ala) by promoting the hydrolysis of Ser-mischarged tRNA(Ala), thereby playing a role in translational fidelity. Binds directly to the catalytic domain of AARS/AlaRS and captures Ser that is misactivated by AARS/AlaRS, preventing the charging of Ser adenylates to tRNA(Ala) and precluding Ser misincorporation in nascent peptides. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANKRD16
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ANKRD16
Gene Accession No. Q6P6B7
Gene Alias 2810455F06Rik; AI646698; Ankrd16; ankyrin repeat domain 16; ankyrin repeat domain-containing protein 16; D430029B21Rik; hypothetical protein LOC550225; si:dkey-108d22.7; Unknown (protein for MGC:143369); zgc:110558
Gene Symbols ANKRD16
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ANKRD16. Recombinant protein control fragment (Product #RP-102793).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54522
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.