Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ANKRD17 Antibody, DyLight 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10086996V
Description
ANKRD17 Polyclonal antibody specifically detects ANKRD17 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
ANKRD17 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
ankyrin repeat domain 17, ankyrin repeat domain-containing protein 17, Gene trap ankyrin repeat protein, GTARSerologically defined breast cancer antigen NY-BR-16, KIAA0697FLJ22206, NY-BR-16 | |
The immunogen recognized by this antibody maps to a region between residue 1550 and 1600 of human ankyrin repeat domain 17 using the numbering given in entry NP_115593.3 (GeneID 26057). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 405 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
26057 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction