Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ANKRD17 Antibody, DyLight 594, Novus Biologicals™
SDP

Catalog No. NB245565 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB245565 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB245565 Supplier Novus Biologicals Supplier No. NB10086996DL594
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ANKRD17 Polyclonal antibody specifically detects ANKRD17 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen ANKRD17
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 594
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias ankyrin repeat domain 17, ankyrin repeat domain-containing protein 17, Gene trap ankyrin repeat protein, GTARSerologically defined breast cancer antigen NY-BR-16, KIAA0697FLJ22206, NY-BR-16
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 1550 and 1600 of human ankyrin repeat domain 17 using the numbering given in entry NP_115593.3 (GeneID 26057).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26057
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.