Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ANKRD17 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560900
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560900 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560900 Supplier Invitrogen™ Supplier No. PA560900
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SSSGSSSAHS NQQQPPGSVS QEPRPPLQQS QVPPPEVRMT VPPLATSSAP VAVPSTAPVT YPMPQTPMGC PQPTPKMETP AIRPPPHGTT APHKN Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 86%.

This gene encodes a protein with ankyrin repeats, which are associated with protein-protein interactions. Studies in mice suggest that this protein is involved in liver development. Three transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANKRD17
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ANKRD17
Gene Accession No. O75179
Gene Alias 4933425K22Rik; A130069E23Rik; A930008M01; AA407558; AA516750; Ankrd17; ankyrin repeat domain 17; ankyrin repeat domain containing protein FOE; ankyrin repeat domain protein 17; ankyrin repeat domain-containing protein 17; Ankyrin repeat domain-containing protein FOE; AU040470; Foe; Gene trap ankyrin repeat protein; Gtar; Kiaa0697; Mask; MASK2; mKIAA0697; NY-BR-16; RGD1562348; serologically defined breast cancer antigen NY-BR-16
Gene Symbols ANKRD17
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ANKRD17. Recombinant protein control fragment (Product #RP-100391).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26057
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.