Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ANKRD17 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS9295R

Description
This gene encodes a protein with ankyrin repeats, which are associated with protein-protein interactions. Studies in mice suggest that this protein is involved in liver development. Three transcript variants encoding different isoforms have been found for this gene.Specifications
ANKRD17 | |
Polyclonal | |
Unconjugated | |
ANKRD17 | |
ANKRD17; ankyrin repeat domain 17; Ankyrin repeat domain-containing protein 17; Gene trap ankyrin repeat protein; GTAR; KIAA0697; MASK2; NY-BR-16; Serologically defined breast cancer antigen NY-BR-16 | |
Rabbit | |
Protein A | |
RUO | |
26057 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin) | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
O75179 | |
ANKRD17 | |
KLH conjugated synthetic peptide derived from human ANKRD17. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction