Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ANKRD18A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546359
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546359 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546359 Supplier Invitrogen™ Supplier No. PA546359
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: MRNVQEKCEK LEKDKKMLEE EVLNLKTHME KDMVELGKLQ EYKSELDERA Sequence homology: Dog: 90%; Horse: 75%; Human: 100%.

The function of this protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANKRD18A
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ANKRD18A
Gene Accession No. Q8IVF6
Gene Alias ANKRD18A; ankyrin repeat domain 18A; ankyrin repeat domain-containing protein 18A; KIAA2015
Gene Symbols ANKRD18A
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human ANKRD18A (aa 781-830).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 253650
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.