Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ANKRD2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA542202
Description
Peptide sequence: QEEENEKLRG DARQKLPMDL LVLEDEKHHG AQSAALQKVK GQERVRKTSL Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
ANKRD2 may play an important role in skeletal muscle hypertrophy.
Specifications
| ANKRD2 | |
| Polyclonal | |
| Unconjugated | |
| Ankrd2 | |
| Ankrd2; ankyrin repeat domain 2; ankyrin repeat domain 2 (stretch responsive muscle); ankyrin repeat domain-containing protein 2; ankyrin repeat domain-containing protein 2; LOW QUALITY PROTEIN: ankyrin repeat domain-containing protein 2; ARPP; hArpp; LOW QUALITY PROTEIN: ankyrin repeat domain-containing protein 2; mArpp; Skeletal muscle ankyrin repeat protein | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 26287 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9GZV1 | |
| Ankrd2 | |
| Synthetic peptide directed towards the N-terminal of human ANKRD2 (aa 51-100). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction