Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ANKRD22 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA571839
Description
ANKRD22 Polyclonal Antibody for Western Blot, IHC (P)
ANKRD22 is a protein coding gene.
Specifications
ANKRD22 | |
Polyclonal | |
Unconjugated | |
ANKRD22 | |
5430429D21Rik; Ankrd22; ankyrin repeat domain 22; ankyrin repeat domain-containing protein 22; D19Ertd675e | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
118932, 52024 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q5VYY1, Q9D3J5 | |
ANKRD22 | |
KLH conjugated synthetic peptide between 140-169 amino acids from the C-terminal region of human ANKRD22. | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction